4-Methoxyphenyl-3-O-allyl-2-azido-4,6-O-benzylidene-2-deoxy-β-D-glucopyranoside ≥98.0% (by HPLC)

Leverandør: TCI

M1638-1G
TCIAM1638-1GEA 3000 NOK
TCIAM1638-1G
4-Methoxyphenyl-3-O-allyl-2-azido-4,6-O-benzylidene-2-deoxy-β-D-glucopyranoside ≥98.0% (by HPLC)
4-Methoxyphenyl-3-O-allyl-2-azido-4,6-O-benzylidene-2-deoxy-beta-D-glucopyranoside
Formel: C₂₃H₂₅N₃O₆
MW: 439,47 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD06797147
CAS-nummer: 889453-78-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 119.0 to 123.0 deg-C
Specific rotation [a]20/D -30.0 to -32.5 deg (C=1, CHCl3)
Elemental analysis (Nitrogen) 9.10 to 10.10 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR