Methyl quinoline-6-carboxylate ≥97.0% (by GC, titration analysis)

Leverandør: TCI

M1687-25G M1687-5G
TCIAM1687-25GEA 2150 NOK
TCIAM1687-25G TCIAM1687-5G
Methyl quinoline-6-carboxylate ≥97.0% (by GC, titration analysis)
Methyl quinoline-6-carboxylate
Formel: C₁₁H₉NO₂
MW: 187.2000 g/mol
MDL nummer: MFCD00956766
CAS-nummer: 38896-30-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8d149942c96f4d574de65e8281b1fc4dd2e5cac&vid=b8d149942c96f4d574de65e8281b1fc4dd2e5cac&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 84.0 to 88.0 deg-C

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR