2-Merkaptoetanol ≥99.0% (by GC) for electrophoresis

Leverandør: TCI
Danger

Synonymer: 2-Hydroxyethyl mercaptan, β-Mercaptoethanol

M1948-25G M1948-5G
TCIAM1948-25GEA 395 NOK
TCIAM1948-25G TCIAM1948-5G
2-Merkaptoetanol ≥99.0% (by GC) for electrophoresis
2-Merkaptoetanol
Formel: HSCH₂CH₂OH
Kokepunkt: 158°C
Smeltepunkt: –100°C
Tetthet: 1,12 g/cm³ (20 °C)
Flash punkt: 74°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004890
CAS-nummer: 60-24-2
EINECS: 200-464-6
FN: 2966
ADR: 6.1,II
Merck Indeks: 14,05869
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9bc34059936db0aeb8f2132e89a4c810b1b642d6&vid=8ac36b8922693d83965d30d9e71e4a80d37e34d8&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless clear liquid
Purity (GC) min. 99.0 %
Refractive index n20/D 1.5000 to 1.5040
Functionality test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR