N-Methylpropylamine ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: N-Methyl-1-propylamine

M2232-5G
TCIAM2232-5GEA 1575 NOK
TCIAM2232-5G
N-Methylpropylamine ≥98.0% (by GC)
N-Methylpropylamine
Formel: C₄H₁₁N
MW: 73.14 g/mol
Kokepunkt: 63°C
Flash punkt: –32°C
MDL nummer: MFCD00009361
CAS-nummer: 627-35-0
FN: 2733
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bab028bffed3c2150fdb02d4538ff712945f799&vid=0bab028bffed3c2150fdb02d4538ff712945f799&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR