Meropenem trihydrate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

M2279-1G M2279-5G
TCIAM2279-1GEA 1050 NOK
TCIAM2279-1G TCIAM2279-5G
Meropenem trihydrate ≥98.0% (by HPLC, titration analysis)
Meropenem trihydrate
Formel: C₁₇H₃₁N₃O₈S
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD08600005
CAS-nummer: 119478-56-7
Merck Indeks: 14,05900

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D -17.0 to -21.0 deg (C=0.44, H2O)
Water 11.4 to 13.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR