Methyl cis-6-octadecenoate 98,0 GC_ASSAY_METHOD

Leverandør: TCI

Synonymer: Petroselinic Acid Methyl Ester, cis-6-Octadecenoic Acid Methyl Ester, Methyl Petroselinate

M2306-500MG
TCIAM2306-500MGEA 1700 NOK
TCIAM2306-500MG
Methyl cis-6-octadecenoate 98,0 GC_ASSAY_METHOD
Methyl cis-6-octadecenoate
Formel: C₁₉H₃₆O₂
MW: 296.50 g/mol
Kokepunkt: 160°C
Smeltepunkt: –1°C
Tetthet: 0.88
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00056193
CAS-nummer: 2777-58-4
EINECS: 220-470-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66ff95084f62e0c01470c8c3d16c741e1f8a3efb&vid=66ff95084f62e0c01470c8c3d16c741e1f8a3efb&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR