Methyl cis-11-octadecenoate ≥98.0% (by GC)

Leverandør: TCI

M2308-100MG
TCIAM2308-100MGEA 960 NOK
TCIAM2308-100MG
Methyl cis-11-octadecenoate ≥98.0% (by GC)
Methyl cis-11-octadecenoate
Formel: C₁₉H₃₆O₂
Kokepunkt: 136°C
Tetthet: 0,88
Flash punkt: 62°C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00010457
CAS-nummer: 1937-63-9
EINECS: 217-714-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9c1d6ebd99d7d4a50b0d54c7908a2b7c19d279f&vid=f9c1d6ebd99d7d4a50b0d54c7908a2b7c19d279f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR