(S)-3-[1-(4-Methoxybenzenesulphonyl)-(4S,5S)-4,5-diphenyl-4,5-dihydro-1H-imidazol-2-yl]-1,1'-binaphthalene-2,2'-diyl Hydrogen Phosphate ≥96.0% (by HPLC)

Leverandør: TCI

M2765-50MG
TCIAM2765-50MGEA 3475 NOK
TCIAM2765-50MG
(S)-3-[1-(4-Methoxybenzenesulphonyl)-(4S,5S)-4,5-diphenyl-4,5-dihydro-1H-imidazol-2-yl]-1,1'-binaphthalene-2,2'-diyl Hydrogen Phosphate ≥96.0% (by HPLC)
(S)-3-[1-(4-Methoxybenzenesulphonyl)-(4S,5S)-4,5-diphenyl-4,5-dihydro-1H-imidazol-2-yl]-1,1'-binaphthalene-2,2'-diyl Hydrogen Phosphate
Formel: C₄₂H₃₁N₂O₇PS
Lagringstemperatur: Fryseskap
CAS-nummer: 1621994-95-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c22821f7791877979c41743d2cbd4341c1e9af59&vid=c22821f7791877979c41743d2cbd4341c1e9af59&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 96.0 area%
Elemental analysis (Nitrogen) 3.50 to 4.00 %
Specific rotation [a]20/D +275 to +295 deg (C=0.5, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR