Methylphosphonic acid ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Danger

M2938-1G
TCIAM2938-1GEA 310 NOK
TCIAM2938-1G
Methylphosphonic acid ≥98.0% (by titrimetric analysis)
Methylphosphonic acid
Formel: CH₅O₃P
MDL nummer: MFCD00002137
CAS-nummer: 993-13-5
EINECS: 213-607-2
FN: 3261
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 106.0 to 110.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR