1,2-Naphthoquinone-4-sulphonic acid sodium salt ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: Folin's reagent, Sodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulphonate

N0043-1G N0043-25G
TCIAN0043-1GEA 220 NOK
TCIAN0043-1G TCIAN0043-25G
1,2-Naphthoquinone-4-sulphonic acid sodium salt ≥98.0% (by titrimetric analysis)
1,2-Naphthoquinone-4-sulphonic acid sodium salt
Formel: C₁₀H₅NaO₅S
MW: 260.20 g/mol
MDL nummer: MFCD00001700
CAS-nummer: 521-24-4
EINECS: 208-308-9
Merck Indeks: 14,08646
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7659c69281bea93dad840b96213013d2b08b8cd&vid=b7659c69281bea93dad840b96213013d2b08b8cd&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (Ion exchange titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR