1-Naphthylamine hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Danger

N0056-25G
TCIAN0056-25GEA 1775 NOK
TCIAN0056-25G
1-Naphthylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
1-Naphthylamine hydrochloride
Formel: C₁₀H₁₀ClN
MDL nummer: MFCD00036370
CAS-nummer: 552-46-5
EINECS: 209-014-3
FN: 2811
ADR: 6.1,III
Merck Indeks: 14,06398

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Dark green powder to crystal
Purity (Neutralization titration) min. 98.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR