Nicotinic acid hydrazide ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: Nicotinohydrazide, (3-Pyridylcarbonyl)hydrazine, Pyridine-3-carbohydrazide

N0087-25G N0087-100G
TCIAN0087-25GEA 285 NOK
TCIAN0087-25G TCIAN0087-100G
Nicotinic acid hydrazide ≥98.0% (by GC, titration analysis)
Nicotinic acid hydrazide
Formel: C₆H₇N₃O
MW: 137.14 g/mol
MDL nummer: MFCD00006383
CAS-nummer: 553-53-7
EINECS: 209-041-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd288a6b350d31d70c3e16a3c4fc526de062d0ac&vid=cd288a6b350d31d70c3e16a3c4fc526de062d0ac&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Bromination Method) min. 98.0 %
Melting point 160.0 to 164.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR