syn-3-Nitrobenzaldoxime ≥98.0% (by GC)

Leverandør: TCI
Warning

N0132-25G
TCIAN0132-25GEA 1450 NOK
TCIAN0132-25G
syn-3-Nitrobenzaldoxime ≥98.0% (by GC)
syn-3-Nitrobenzaldoxime
Formel: C₇H₆N₂O₃
MW: 166.14 g/mol
MDL nummer: MFCD00007275
CAS-nummer: 3717-29-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 119.0 to 122.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR