5-Nitro-1,10-phenanthroline ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

N0214-1G
TCIAN0214-1GEA 420 NOK
TCIAN0214-1G
5-Nitro-1,10-phenanthroline ≥98.0% (by HPLC, titration analysis)
5-Nitro-1,10-phenanthroline
Formel: C₁₂H₇N₃O₂
Smeltepunkt: 203 °C
MDL nummer: MFCD00004981
CAS-nummer: 4199-88-6
EINECS: 224-097-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1517b27eff45e30d545b813d089146577fd85688&vid=1517b27eff45e30d545b813d089146577fd85688&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 200.0 to 203.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR