5-Nitro-2-furaldehyde diacetate ≥97.0% (by GC)

Leverandør: TCI

Synonymer: 5-Nitrofurfural Diacetyl Acetal, Nitrofurfural diacetate, 2-(Diacetoxymethyl)-5-nitrofuran, (5-Nitrofuran-2-yl)methylene diacetate, 5-Nitrofurfurylidene Diacetate

N0218-25G
TCIAN0218-25GEA 1550 NOK
TCIAN0218-25G
5-Nitro-2-furaldehyde diacetate ≥97.0% (by GC)
5-Nitro-2-furaldehyde diacetate
Formel: C₉H₉NO₇
MDL nummer: MFCD00003244
CAS-nummer: 92-55-7
EINECS: 202-166-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 87.0 to 92.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR