Nalidixic acid ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Danger

N0490-25G
TCIAN0490-25GEA 930 NOK
TCIAN0490-25G
Nalidixic acid ≥95.0% (by titrimetric analysis)
Nalidixic acid
Formel: C₁₂H₁₂N₂O₃
MW: 232,24 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006884
CAS-nummer: 389-08-2
EINECS: 206-864-7
Merck Indeks: 14,06359
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99c4a1043ae287287916f90bbf026a0f68c03ac4&vid=99c4a1043ae287287916f90bbf026a0f68c03ac4&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 %
Melting point 227.0 to 231.0 deg-C
Solubility in Sodium hydroxide solution very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR