Nicardipine hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Danger

Synonymer: 3-(2-(Benzyl(methyl)amino)ethyl) 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate hydrochloride, Nicardipine HCl

N0635-5G
TCIAN0635-5GEA 1275 NOK
TCIAN0635-5G
Nicardipine hydrochloride ≥98.0% (by HPLC, titration analysis)
Nicardipine hydrochloride
Formel: C₂₆H₂₉N₃O₆
MW: 515,99 g/mol
MDL nummer: MFCD00057327
CAS-nummer: 54527-84-3
EINECS: 259-198-4
FN: 2811
ADR: 6.1,III
Merck Indeks: 14,06495

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR