5-Nitroindole-2-carboxylic acid

Leverandør: TCI

Synonymer: 5-Nitro-1H-indole-2-carboxylic acid

N1036-1G
TCIAN1036-1GEA 2200 NOK
TCIAN1036-1G
5-Nitroindole-2-carboxylic acid
5-Nitroindole-2-carboxylic acid
Formel: C₉H₆N₂O₄
MW: 206,157 g/mol
MDL nummer: MFCD00236372
CAS-nummer: 16730-20-4

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR