Neohesperidin ≥97.0% (by HPLC)

Leverandør: TCI

Synonymer: (S)-7-(((2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)chroman-4-one, (S)-4′-Methoxy-3′,5,7-trihydroxyflavanone-7-[2-O-(α-L-rhamnopyranosyl)-β-D-glucopyranoside]

N1121-100MG
TCIAN1121-100MGEA 1650 NOK
TCIAN1121-100MG
Neohesperidin ≥97.0% (by HPLC)
Neohesperidin
Formel: C₂₈H₃₄O₁₅
MW: 610,60 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD16661259
CAS-nummer: 13241-33-3
EINECS: 236-216-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Melting point 241.0 to 245.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR