n-Oktan ≥98.5% (by GC) for HPLC

Leverandør: TCI
Danger

O0151-500ML
TCIAO0151-500MLEA 16525 NOK
TCIAO0151-500ML
n-Oktan ≥98.5% (by GC) for HPLC
n-Oktan
Formel: H₃C(CH₂)₆CH₃
Kokepunkt: 125°C
Smeltepunkt: –57°C
Tetthet: 0,703 g/cm³ (20 °C)
Flash punkt: 14°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009556
CAS-nummer: 111-65-9
EINECS: 203-892-1
FN: 1262
ADR: 3,II
Merck Indeks: 14,06749
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6370c5f2c5ee9957a468cb919de3892bcb8a283&vid=b506148c950c3881365d43f3a14206ee01c9c87c&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless clear liquid
Purity (GC) min. 98.5 %
Absorbance (210nm) max. 0.350
Absorbance (220nm) max. 0.150
Absorbance (230nm) max. 0.060


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR