Octadecylbenzene ≥95.0% (by GC)

Leverandør: TCI

Synonymer: 1-Phenyloctadecane

O0160-25G O0160-5G
TCIAO0160-25GEA 2775 NOK
TCIAO0160-25G TCIAO0160-5G
Octadecylbenzene ≥95.0% (by GC)
Octadecylbenzene
Formel: C₂₄H₄₂
Kokepunkt: 408 °C (1013 hPa)
Smeltepunkt: 29 °C
Tetthet: 0,8546 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00048500
CAS-nummer: 4445-07-2
EINECS: 224-684-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78ee05e130a0b1b1c1c3116ab5b7bc3bf2b49189&vid=164d0e34ccb86413c7e4531b9b810763de62ea46&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 95.0 %
Freezing point 32.0 to 36.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR