1,8-Diacetoxyoctane

Leverandør: TCI

Synonymer: Octamethylene Diacetate, 1,8-Octanediol Diacetate

O0275-25G
TCIAO0275-25GEA 890 NOK
TCIAO0275-25G
1,8-Diacetoxyoctane
1,8-Diacetoxyoctane
Formel: C₁₂H₂₂O₄
Kokepunkt: 128°C
MDL nummer: MFCD01861294
CAS-nummer: 3992-48-1

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR