Olopatadine hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: (Z)-2-(11-(3-(Dimethylamino)propylidene)-6,11-dihydrodibenzo[b,e]oxepin-2-yl)acetic acid hydrochloride, Olopatadine HCl

O0361-1G
TCIAO0361-1GEA 2450 NOK
TCIAO0361-1G
Olopatadine hydrochloride ≥98.0% (by HPLC, titration analysis)
Olopatadine hydrochloride
Formel: C₂₁H₂₃NO₃
MW: 373,88 g/mol
MDL nummer: MFCD00875716
CAS-nummer: 140462-76-6
Merck Indeks: 14,06840

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) 98.0 to 102.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR