2-Oxa-7-azaspiro[3.5]nonane hemioxalate ≥97.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: 2-Oxa-7-azaspiro[3.5]nonane oxalate(2:1)

O0509-250MG
TCIAO0509-250MGEA 1525 NOK
TCIAO0509-250MG
2-Oxa-7-azaspiro[3.5]nonane hemioxalate ≥97.0% (by titrimetric analysis)
2-Oxa-7-azaspiro[3.5]nonane hemioxalate
Formel: C₁₆H₂₈N₂O₆
MW: 344,4 g/mol
MDL nummer: MFCD11035888
CAS-nummer: 1429056-28-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 97.0 % (calcd.on free amine, dried substance)
Oxalic Acid 25.0 to 44.0 %
Water max. 3.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR