Octadecyl rhodamine B chloride

Leverandør: TCI
Warning

Synonymer: 6-(Diethylamino)-N,N-diethyl-9-[2-[(octadecyloxy)carbonyl]phenyl]-3H-xanthen-3-iminium chloride, 3,6-Bis(diethylamino)-9-[2-[(octadecyloxy)carbonyl]phenyl]xanthylium chloride, N-[6-(Diethylamino)-9-[2-[(octadecyloxy)carbonyl]phenyl]-3H-xanthen-3-ylidene]-N-ethylethanaminium chloride

O0512-10MG O0512-50MG
TCIAO0512-10MGEA 1900 NOK
TCIAO0512-10MG TCIAO0512-50MG
Octadecyl rhodamine B chloride
Octadecyl rhodamine B chloride
Formel: C₄₆H₆₇ClN₂O₃
MDL nummer: MFCD00467892
CAS-nummer: 65603-19-2

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Amber to Dark red powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) gt. 3.5 to 4.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR