Organosilica Rhodamine B, Ø 100 nm

Leverandør: TCI

O0573-2MG
TCIAO0573-2MGEA 10725 NOK
TCIAO0573-2MG
Organosilica Rhodamine B, Ø 100 nm
Organosilica Rhodamine B

For use as a fluorophore in the fluorescent labeling of proteins and other molecules. Incorporation of Rhodamine B fluorescent dye into conjugatable silica nanoparticles allows for superior fluorescence intensity and light resistance when compared to unmodified Rhodamine B. Thiol groups (-SH) at the surface of the nanoparticles provide a convenient mechanism for conjugation. This product is made without toxic metals.


Product Information
Volume/Quantity : 2 mg
Product Form : Suspension in water
Functional Group : Thiol group (-SH)
Excitation / Emission : 556 nm / 579 nm


NB: Store at 4 °C. Do not freeze.

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR