Fenol rødt

Leverandør: TCI
Warning

P0100-1G P0100-25G P0100-100G
TCIAP0100-1GEA 220 NOK
TCIAP0100-1G TCIAP0100-25G TCIAP0100-100G
Fenol rødt
Fenol rødt
Formel: C₁₉H₁₄O₅S
Smeltepunkt: > 300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003552
CAS-nummer: 143-74-8
EINECS: 205-609-7
Merck Indeks: 14,07247
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e51da0f259217a65cf4d8881c4c62bba3ec1269&vid=6e51da0f259217a65cf4d8881c4c62bba3ec1269&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Dark red to Brown powder to crystaline
Absorbance min. 0.530 (5mg/L, pH 8.4, 556.0 to 560.0 nm)
Transition interval (pH) (yellow)6.8 to 8.4 (red)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR