trans-Cinnamoyl chloride ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: trans-3-Phenylacryloyl Chloride, (E)-Cinnamoyl chloride

P0133-25G
TCIAP0133-25GEA 325 NOK
TCIAP0133-25G
trans-Cinnamoyl chloride ≥97.0% (by titrimetric analysis)
trans-Cinnamoyl chloride
Formel: C₉H₇ClO
MW: 166,61 g/mol
MDL nummer: MFCD00000732
CAS-nummer: 17082-09-6
EINECS: 203-065-5
FN: 3261
ADR: 8,III
Merck Indeks: 14,02301
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83c22e6647a35ffb80bc70062aec937c48b64b51&vid=83c22e6647a35ffb80bc70062aec937c48b64b51&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 95.0 %
Purity (Argentmetric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR