Phenylhydrazine sulphate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Danger

P0186-25G
TCIAP0186-25GEA 865 NOK
TCIAP0186-25G
Phenylhydrazine sulphate ≥98.0% (by titrimetric analysis)
Phenylhydrazine sulphate
Formel: C₆H₈N₂
MDL nummer: MFCD00070608
CAS-nummer: 52033-74-6
EINECS: 257-622-2
FN: 2811
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (Potassium iodate Method) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR