Cresyl diphenyl phosphate (mixture of analogue) so_called 93,0 GC_ASSAY_METHOD

Leverandør: TCI
Warning

Synonymer: Diphenyl p-tolyl phosphate, Diphenyl Cresyl Phosphate, Phosphoric Acid Cresyl Diphenyl Ester

P0259-25ML P0259-500ML
TCIAP0259-25MLEA 230 NOK
TCIAP0259-25ML TCIAP0259-500ML
Cresyl diphenyl phosphate (mixture of analogue) so_called 93,0 GC_ASSAY_METHOD
Cresyl diphenyl phosphate (mixture of analogue)
Formel: C₁₉H₁₇O₄P
MW: 340.3097 g/mol
Kokepunkt: 390°C
Flash punkt: 232°C
MDL nummer: MFCD00014920
CAS-nummer: 26444-49-5
EINECS: 247-693-8
FN: 3082
ADR: 9 III

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 93.0 % (total of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR