2-Ethylhexyl phosphate (mono and di ester mixture)

Supplier: TCI
Danger

P0261-25G P0261-500G
TCIAP0261-25GEA 220 NOK
TCIAP0261-25G TCIAP0261-500G
2-Ethylhexyl phosphate (mono and di ester mixture)
2-Ethylhexyl phosphate (mono and di ester mixture)
MDL Number: MFCD00135884
CAS Number: 12645-31-7
EINECS: 235-741-0
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Assay of mono ester 30.0 to 45.0 %
Assay of Diester 50.0 to 70.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR