4-(Isopropylamino)diphenylamine ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: N1-Isopropyl-N4-phenylbenzene-1,4-diamine, N-Isopropyl-N'-phenyl-p-phenylenediamine

P0327-25G P0327-500G
TCIAP0327-25GEA 220 NOK
TCIAP0327-25G TCIAP0327-500G
4-(Isopropylamino)diphenylamine ≥98.0% (by HPLC, titration analysis)
4-(Isopropylamino)diphenylamine
Formel: C₁₅H₁₈N₂
Kokepunkt: 180°C
Flash punkt: 198°C
MDL nummer: MFCD00026348
CAS-nummer: 101-72-4
EINECS: 202-969-7
FN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16c0ae92b586f0676df17dc4d8cb94c58c1b564a&vid=16c0ae92b586f0676df17dc4d8cb94c58c1b564a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 77.0 to 82.0 deg-C

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR