Fenantren ≥98.5%, zone refined: number of passes 30

Leverandør: TCI
Warning

P0331-1SAMPLE
TCIAP0331-1SAMPLEEA 4800 NOK
TCIAP0331-1SAMPLE
Fenantren ≥98.5%, zone refined: number of passes 30
Fenantren

CAS Number: 85-01-8
MDL Number: MFCD00001168
Molecular Formula: C14H10
Molecular Weight: 178.23
Purity/Analysis Method: >98.5% (GC)
Form: Crystal
Boiling point (°C): 340
Melting point (°C): 100
Flash Point (°C): 171

Formel: C₁₄H₁₀
Kokepunkt: 340 °C
Smeltepunkt: 100 °C
Tetthet: 1,19 g/cm³ (20 °C)
Flash punkt: 171 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001168
CAS-nummer: 85-01-8
EINECS: 201-581-5
FN: 3077
ADR: 9,III
Merck Indeks: 12,07354

Bestill nå

Spesifikasjon Testresultater

Purity/Analysis Method >98.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR