Poly(ethylene glycol succinate)

Leverandør: TCI

Synonymer: Ethylene Glycol Succinate Polyester, Succinate Ethylene Glycol Polyester, Poly(ethylene Glycol Succinate)

P0467-25G
TCIAP0467-25GEA 1600 NOK
TCIAP0467-25G
Poly(ethylene glycol succinate)
Poly(ethylene glycol succinate)
Formel: (C₄H₆O₄·C₂H₆O₂)x
MDL nummer: MFCD00084421
CAS-nummer: 25569-53-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Suitability for GC-analysis to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR