Palatine chrome green

Leverandør: TCI

Synonymer: Mordant Green 17

P0594-25G
TCIAP0594-25GEA 875 NOK
TCIAP0594-25G
Palatine chrome green
Palatine chrome green
Formel: C₁₆H₁₂N₄O₁₀S₂
MW: 528,37 g/mol
MDL nummer: MFCD00135102
CAS-nummer: 3564-28-1
EINECS: 222-646-4

Bestill nå

Spesifikasjon Testresultater

Appearance Black powder to crystal
Lambda max. 540.0 to 560.0 nm (H2O)
Absorbance (E1%1cm) min. 300 (H2O, 540.0 to 560.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR