Acid blue 1

Leverandør: TCI

Synonymer: Alphazurine G, Patent Blue V, Carmine Blue V, Xylene Blue, Brilliant Acid Blue, Azure Blue VX

P0597-25G
TCIAP0597-25GEA 635 NOK
TCIAP0597-25G
Acid blue 1
Acid blue 1
Formel: C₂₇H₃₂N₂O₆S₂
MW: 544.68 g/mol
MDL nummer: MFCD00012119
CAS-nummer: 116-95-0
EINECS: 204-165-1
Merck Indeks: 14,08987

Bestill nå

Spesifikasjon Testresultater

Appearance Yellow to Brown to Dark green powder to crystal
Lambda max. 637.0 to 641.0 nm (H2O)
Absorbance min. 0.800 (4mg/L, H2O, 637.0 to 641.0 nm) (calcd.on dried substance)
Drying loss max. 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR