Sodium pyridine-3-sulphonate ≥97.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: Pyridine-3-sulphonic acid Sodium Salt

P0618-25G
TCIAP0618-25GEA 1225 NOK
TCIAP0618-25G
Sodium pyridine-3-sulphonate ≥97.0% (by HPLC, titration analysis)
Sodium pyridine-3-sulphonate
Formel: C₅H₅NO₃S
MW: 181,14 g/mol
MDL nummer: MFCD00065041
CAS-nummer: 15521-77-4
EINECS: 239-561-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR