Perfluorokerosene, boiling_low Mass spectrometry

Leverandør: TCI
Warning

Synonymer: PFK

P0690-10G
TCIAP0690-10GEA 1725 NOK
TCIAP0690-10G
Perfluorokerosene, boiling_low Mass spectrometry
Perfluorokerosene
Flash punkt: 46°C
MDL nummer: MFCD00132835
FN: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=116a12ead0ed56c97bbd63234ef1670cdacb369d&vid=ff76e9f219e6c63a94ceb39d9149d15c1dbaf358&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Suitability for Mass-analysis calibratio to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR