Pentaerythritol distearate (so called) mixture ≥95.0% (by titrimetric analysis)

Leverandør: TCI

P0738-25G
TCIAP0738-25GEA 525 NOK
TCIAP0738-25G
Pentaerythritol distearate (so called) mixture ≥95.0% (by titrimetric analysis)
Pentaerythritol distearate (so called)
Formel: C₄₁H₈₀O₆
MDL nummer: MFCD00059225
CAS-nummer: 13081-97-5
EINECS: 235-991-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light red powder to crystal
Purity (Ester Value) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR