Diphenyl phosphate 99,0 HPLC, T

Leverandør: TCI

Synonymer: Diphenyl hydrogen phosphate

P0801-1G P0801-25G
TCIAP0801-1GEA 220 NOK
TCIAP0801-1G TCIAP0801-25G
Diphenyl phosphate 99,0 HPLC, T
Diphenyl phosphate
Formel: C₁₂H₁₁O₄P
MW: 250.19 g/mol
MDL nummer: MFCD00003033
CAS-nummer: 838-85-7
EINECS: 212-657-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e0d509d4fc95c495eed741b507405ca01cbd340&vid=1e0d509d4fc95c495eed741b507405ca01cbd340&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 99.0 %
Melting point 67.0 to 71.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR