Bis(2-ethylhexyl)phosphate ≥95.0% (by titrimetric analysis) for rare metals extraction

Leverandør: TCI
Danger

P0811-25ML P0811-500ML
TCIAP0811-25MLEA 230 NOK
TCIAP0811-25ML TCIAP0811-500ML
Bis(2-ethylhexyl)phosphate ≥95.0% (by titrimetric analysis) for rare metals extraction
Bis(2-ethylhexyl)phosphate
Formel: C₁₆H₃₅O₄P
Smeltepunkt: -50°C
Flash punkt: 196°C
MDL nummer: MFCD00009492
CAS-nummer: 298-07-7
EINECS: 206-056-4
FN: 1902
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d72f139b09afa5f0f3ce2e0a74387fe998e106c&vid=0c5ac6429daab9153bdb02e6bdbd69fdd87d2d5f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (Neutralization titration) min. 95.0 %
Specific gravity (20/20) 0.9730 to 0.9830
Refractive index n20/D 1.4420 to 1.4440


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR