2-(Phenylsulphonyl)acetophenone ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: 1-Phenyl-2-(phenylsulphonyl)ethanone

P0828-5G
TCIAP0828-5GEA 1425 NOK
TCIAP0828-5G
2-(Phenylsulphonyl)acetophenone ≥98.0% (by titrimetric analysis)
2-(Phenylsulphonyl)acetophenone
Formel: C₁₄H₁₂O₃S
MW: 260,308 g/mol
MDL nummer: MFCD00025043
CAS-nummer: 3406-03-9
EINECS: 222-292-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=356f64068032fdff688ba11d40c9c8dcb438a90f&vid=356f64068032fdff688ba11d40c9c8dcb438a90f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 91.0 to 95.0 deg-C
Solubility in hot Toluene very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR