Pivalaldehyde (trimethylacetaldehyde) ≥95.0% (by GC)

Leverandør: TCI
Danger

Synonymer: Pivalaldehyde, Pivaldehyde, Trimethylacetaldehyde, 2,2-Dimethylpropionaldehyde, 2,2-Dimethylpropanal

P0847-5ML
TCIAP0847-5MLEA 350 NOK
TCIAP0847-5ML
Pivalaldehyde (trimethylacetaldehyde) ≥95.0% (by GC)
Pivalaldehyde (trimethylacetaldehyde)
Formel: C₅H₁₀O
MW: 86.13 g/mol
Kokepunkt: 75°C
Smeltepunkt: 6
Flash punkt: 20°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00006962
CAS-nummer: 630-19-3
EINECS: 211-134-6
FN: 1989
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2a3258ca834f24655964dc5fd42c75f16224c81&vid=f2a3258ca834f24655964dc5fd42c75f16224c81&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR