1,10-Fenatrolin monohydrat ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: 1,10-Phenanthroline hydrate

P0879-25G
TCIAP0879-25GEA 690 NOK
TCIAP0879-25G
1,10-Fenatrolin monohydrat ≥98.0% (by titrimetric analysis)
1,10-Fenatrolin monohydrat
Formel: C₁₂H₈N₂·1H₂O
MW: 198,22 g/mol
Smeltepunkt: 102°C
Tetthet: 1,259 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149973
CAS-nummer: 5144-89-8
EINECS: 200-629-2
FN: 2811
ADR: 6.1,III
Merck Indeks: 14,07214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=160655fadcff37b08c57c69a14d6ba38368e9a1c&vid=160655fadcff37b08c57c69a14d6ba38368e9a1c&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Dark green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR