cis-3-Hexenyl pyruvate ≥95.0% (by GC, titration analysis)

Leverandør: TCI

Synonymer: Pyruvic Acid cis-3-Hexenyl Ester

P0894-25ML
TCIAP0894-25MLEA 1300 NOK
TCIAP0894-25ML
cis-3-Hexenyl pyruvate ≥95.0% (by GC, titration analysis)
cis-3-Hexenyl pyruvate
Formel: C₉H₁₄O₃
MW: 170,21 g/mol
Kokepunkt: 98°C
Flash punkt: 108°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00036527
CAS-nummer: 68133-76-6
EINECS: 268-703-7

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 95.0 %
Purity (Iodometric Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR