Tris(2-ethylhexyl)phosphate ≥98.0% (by GC)

Leverandør: TCI
Warning

P1022-25ML P1022-500ML
TCIAP1022-25MLEA 245 NOK
TCIAP1022-25ML TCIAP1022-500ML
Tris(2-ethylhexyl)phosphate ≥98.0% (by GC)
Tris(2-ethylhexyl)phosphate
Formel: C₂₄H₅₁O₄P
Kokepunkt: 215°C
Flash punkt: 204°C
MDL nummer: MFCD00009491
CAS-nummer: 78-42-2
EINECS: 201-116-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=267c04cdc182bcfaf0b72b124da3eb883e926b2a&vid=267c04cdc182bcfaf0b72b124da3eb883e926b2a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR