Perfluorotriamylamine (so called)

Leverandør: TCI
Warning

Synonymer: Perfluorotripentylamine

P1051-25G
TCIAP1051-25GEA 795 NOK
TCIAP1051-25G
Perfluorotriamylamine (so called)
Perfluorotriamylamine
Formel: C₁₅F₃₃N
MW: 821,12 g/mol
MDL nummer: MFCD00042367
CAS-nummer: 338-84-1

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) many component mixture
Specific gravity (20/20) 1.9380 to 1.9440

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR