(Perfluorohexyl)phenyliodonium trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: (Tridecafluorohexyl)phenyliodonium Trifluoromethanesulphonate

P1080-1G
TCIAP1080-1GEA 2200 NOK
TCIAP1080-1G
(Perfluorohexyl)phenyliodonium trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
(Perfluorohexyl)phenyliodonium trifluoromethanesulphonate
Formel: C₁₃H₅F₁₆IO₃S
MW: 672,12 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00135119
CAS-nummer: 77758-84-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (Potassium iodate Method) min. 98.0 %
Melting point 120.0 to 126.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR