2-Phenylbenzimidazole ≥98.0% (by GC, titration analysis)

Leverandør: TCI

Synonymer: 2-Phenyl-1H-benzimidazole, 2-Phenyl-1H-benzo[d]imidazole

P1105-5G
TCIAP1105-5GEA 260 NOK
TCIAP1105-5G
2-Phenylbenzimidazole ≥98.0% (by GC, titration analysis)
2-Phenylbenzimidazole
Formel: C₁₃H₁₀N₂
MW: 194,24 g/mol
Kokepunkt: 422 °C (1013 hPa)
Smeltepunkt: 211 °C
MDL nummer: MFCD00005592
CAS-nummer: 716-79-0
EINECS: 211-939-2
Merck Indeks: 14,07274
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29c2d8f51453730c7a88d26ec8c9193991047597&vid=29c2d8f51453730c7a88d26ec8c9193991047597&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR