[2.2]-Paracyclophane ≥99.0% (by GC)

Leverandør: TCI

Synonymer: Tricyclo[8,2,2,24,7]hexadeca-4,6,10,12,13,15-hexaene, Tricyclo[8.2.2.2(4,7)]hexadeca-1(12),4,6,10,13,15-hexaene

P1165-1G
TCIAP1165-1GEA 1150 NOK
TCIAP1165-1G
[2.2]-Paracyclophane ≥99.0% (by GC)
[2.2]-Paracyclophane
Formel: C₁₆H₁₆
MW: 208.3 g/mol
Kokepunkt: 321
MDL nummer: MFCD00003707
CAS-nummer: 1633-22-3
EINECS: 216-644-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70f3307d309fe3e5a3ec9d0d7e152edfe2944eb7&vid=70f3307d309fe3e5a3ec9d0d7e152edfe2944eb7&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR